ADV371425935 AChemBlock

4-Hydroxymethyl-3-methylpyridine

  • MDL number MFCD04038635
  • Cas Number : 38070-73-4
  • Molecular Weight : 123.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid/off-white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-hydroxymethyl-3-methylpyridine.html

Safety Information

signalword warning
pcodes P261;P271;P280
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2894
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart