ADV371425938 AChemBlock

3-Azabicyclo[3.1.0]hexane hydrochloride

  • MDL number MFCD12197361
  • Cas Number : 73799-64-1
  • Molecular Weight : 119.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid/off white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-azabicyclo-3-1-0-hexane-hydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2898
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart