ADV371425940 AChemBlock

Benzyl [2-(3,5-bis(trifluoromethyl)phenyl)-1-methyl-2-oxo-ethyl]carbamate

  • MDL number MFCD19707642
  • Cas Number : 1363382-26-6
  • Molecular Weight : 419.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/benzyl-2-3-5-bis-trifluoromethyl-phenyl-1-methyl-2-oxo-ethyl-carbamate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2903
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart