ADV371425943 AChemBlock

methyl (1-(3,5-bis(trifluoromethyl)phenyl)-1-oxopropan-2-yl)carbamate

  • MDL number MFCD19707643
  • Cas Number : 1363382-18-6
  • Molecular Weight : 343.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/rac-carbamic-acid-n-2-3-5-bis-trifluoromethyl-phenyl-1-methyl-2-oxoethyl-methyl-ester.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2906
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart