ADV371425944 AChemBlock

tert-Butyl 5-(hydroxymethyl)pyridin-2-ylcarbamate

  • MDL number MFCD07186264
  • Cas Number : 169280-83-5
  • Molecular Weight : 224.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-5-hydroxymethyl-pyridin-2-ylcarbamate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2907
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart