ADV371425955 AChemBlock

4-Hydroxy-N-[4-methoxy-7-(4-morpholinyl)-2-benzothiazolyl]-4-methyl-1-piperidinecarboxamide

  • MDL number MFCD21642026
  • Cas Number : 870070-55-6
  • Molecular Weight : 406.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-hydroxy-n-4-methoxy-7-4-morpholinyl-2-benzothiazolyl-4-methyl-1-piperidinecarboxamide.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2920
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart