ADV371425961 AChemBlock

Methyl 5-amino-6-bromo-2-pyrazinecarboxylate

  • MDL number MFCD09840911
  • Cas Number : 1076198-49-6
  • Molecular Weight : 232.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form light-yellow solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/methyl-5-amino-6-bromo-2-pyrazinecarboxylate.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2926
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart