ADV371425964 AChemBlock

6-Boc-4-dichloro-5,6,7,8-tetrahydropyrido[4,3-d]pyrimidine

  • MDL number MFCD08273922
  • Cas Number : 1056934-87-2
  • Molecular Weight : 269.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/6-boc-4-dichloro-5-6-7-8-tetrahydropyrido-4-3-d-pyrimidine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2929
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart