ADV371425971 AChemBlock

Ethyl 7-bromopyrazolo[1,5-a]pyrimidine-3-carboxylate

  • MDL number MFCD19707047
  • Cas Number : 1389302-26-4
  • Molecular Weight : 270.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/ethyl-7-bromopyrazolo-1-5-a-pyrimidine-3-carboxylate.html

Safety Information

signalword warning
pcodes P261;P271;P280
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2938
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart