ADV371425973 AChemBlock

Darapladib

  • MDL number MFCD18251452
  • Cas Number : 356057-34-6
  • Molecular Weight : 666.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Physical Form light-yellow solid/off-white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/darapladib.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2941
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart