ADV371425986 AChemBlock

4-Bromo-6-methyl-2-methoxyphenol

  • MDL number MFCD09263254
  • Cas Number : 86232-54-4
  • Molecular Weight : 217.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-bromo-6-methyl-2-methoxyphenol.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2954
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart