ADV371425987 AChemBlock

2-Chloro-3-hydroxyquinoline-4-carboxylic acid

  • MDL number MFCD11706991
  • Cas Number : 847547-91-5
  • Molecular Weight : 223.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-chloro-3-hydroxyquinoline-4-carboxylic-acid.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2955
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart