ADV371426001 AChemBlock

Pyrrolo[1,2-f][1,2,4]triazin-4-amine

  • MDL number MFCD08234647
  • Cas Number : 159326-68-8
  • Molecular Weight : 134.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form brown solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/pyrrolo-1-2-f-1-2-4-triazin-4-amine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=C-2971
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart