ADV371426034 AChemBlock

3-Cyanopiperidine

  • MDL number MFCD01321091
  • Cas Number : 7492-88-8
  • Molecular Weight : 110.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/3-cyanopiperidine.html

Safety Information

signalword Danger
pcodes P264;P270;P280;P301+312;P305+351+338;P310;P330;P510
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=A-124
pictograms GHS05;GHS07
hstatements H302;H318
Availability details for
Quantity
  • Availability
  • Add to Cart