ADV371426037 AChemBlock

4-(Aminoethyl)-1-N-Boc-piperidine

  • MDL number MFCD04038459
  • Cas Number : 146093-46-1
  • Molecular Weight : 228.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form pale yellow liquid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-aminoethyl-1-n-boc-piperidine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=A-127
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart