ADV371426038 AChemBlock

4-(2-Boc-aminoethyl)piperidine

  • MDL number MFCD01318373
  • Cas Number : 165528-81-4
  • Molecular Weight : 228.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form White Solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-2-boc-aminoethyl-piperidine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=A-128
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart