ADV371426048 AChemBlock

(S)-1-Boc-piperazine-2-carboxylic acid

  • MDL number MFCD02179105
  • Cas Number : 159532-59-9
  • Molecular Weight : 230.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white powder
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/s-1-boc-piperazine-2-carboxylic-acid.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=A-164
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart