ADV371426051 AChemBlock

(R)-1-N-Boc-piperazine-2-carboxylic acid methyl ester

  • MDL number MFCD04115327
  • Cas Number : 252990-05-9
  • Molecular Weight : 244.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form colorless or light-yellow liquid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/r-1-n-boc-piperazine-2-carboxylic-acid-methyl-ester.html

Safety Information

signalword Danger
pcodes P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=A-167
pictograms GHS05; GHS07
hstatements H314;H319
Availability details for
Quantity
  • Availability
  • Add to Cart