ADV371426063 AChemBlock

(S)-2-(Aminomethyl)-1-N-Boc-piperidine

  • MDL number MFCD06799358
  • Cas Number : 475105-35-2
  • Molecular Weight : 214.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/s-2-aminomethyl-1-n-boc-piperidine.html

Safety Information

signalword Danger
pcodes P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=A-179
pictograms GHS05; GHS07
hstatements H314;H318
Availability details for
Quantity
  • Availability
  • Add to Cart