ADV371426088 AChemBlock

N-Methyl-N-((3R,4R)-4-methylpiperidin-3-yl)-7H-pyrrolo[2,3-d]pyrimidin-4-amine

  • MDL number MFCD09878608
  • Cas Number : 477600-74-1
  • Molecular Weight : 245.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/n-methyl-n-3r-4r-4-methylpiperidin-3-yl-7h-pyrrolo-2-3-d-pyrimidin-4-amine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=A-524
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart