ADV371426091 AChemBlock

3-Aza-bicyclo[3.1.1]heptan-6-ol hydrochloride

  • MDL number MFCD22415249
  • Cas Number : 1389264-28-1
  • Molecular Weight : 149.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid/yellow solid/off white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-aza-bicyclo-3-1-1-heptan-6-ol-hydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=F-4015
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart