ADV371426144 AChemBlock

(R)-1-Boc-pyrrolidine-3-carboxylic acid methyl ester

  • MDL number MFCD09954284
  • Cas Number : 441717-40-4
  • Molecular Weight : 229.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/r-1-boc-pyrrolidine-3-carboxylic-acid-methyl-ester.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=A-321
pictograms GHS07
hstatements H313;H303;H333
Availability details for
Quantity
  • Availability
  • Add to Cart