ADV371426152 AChemBlock

(S)-tert-Butyl 3-((methylamino)methyl)pyrrolidine-1-carboxylate

  • MDL number MFCD11616684
  • Cas Number : 1245642-79-8
  • Molecular Weight : 214.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form colorless liquid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/s-tert-butyl-3-methylamino-methyl-pyrrolidine-1-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=A-351
pictograms GHS07
hstatements H313;H303;H333
Availability details for
Quantity
  • Availability
  • Add to Cart