ADV371426159 AChemBlock

(2R,4S)-1-tert-Butyl 2-methyl 4-aminopyrrolidine-1,2-dicarboxylate

  • MDL number MFCD08704545
  • Cas Number : 254881-77-1
  • Molecular Weight : 244.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/2r-4s-1-tert-butyl-2-methyl-4-aminopyrrolidine-1-2-dicarboxylate.html

Safety Information

signalword warning
pcodes P261;P264;P280;P301+312;P302+P352;
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=A-361
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart