ADV371426163 AChemBlock

(S)-2-Cyanopyrrolidine hydrochloride

  • MDL number MFCD02683060
  • Cas Number : 65732-69-6
  • Molecular Weight : 132.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Store at Room Tem.
Physical Form white powder
Shipping Temperature Store at Room Tem.
Country of Origin CN
Product Link https://www.achemblock.com/s-2-cyanopyrrolidine-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=A-367
pictograms GHS07
hstatements H313;H303;H333
Availability details for
Quantity
  • Availability
  • Add to Cart