ADV371426166 AChemBlock

1-Boc-3-aminoazetidine

  • MDL number MFCD01861753
  • Cas Number : 193269-78-2
  • Molecular Weight : 172.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form colorless liquid/light-yellow liquid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-boc-3-aminoazetidine.html

Safety Information

signalword Danger
pcodes P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=A-400
pictograms GHS05
hstatements H314;H318
Availability details for
Quantity
  • Availability
  • Add to Cart