ADV371426194 AChemBlock

3-Hydroxy-cyclobutanecarboxylic acid ethyl ester

  • MDL number MFCD09758972
  • Cas Number : 17205-02-6
  • Molecular Weight : 144.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form colorless or light-yellow liquid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/3-hydroxy-cyclobutanecarboxylic-acid-ethyl-ester.html

Safety Information

signalword Warning
pcodes P210;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=A-432
pictograms GHS02
hstatements H226;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart