ADV371426224 AChemBlock

5-(Trifluoromethyl)-2,3-dihydroinden-1-one

  • MDL number MFCD07772122
  • Cas Number : 150969-56-5
  • Molecular Weight : 200.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form yellow solid/white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-trifluoromethyl-2-3-dihydroinden-1-one.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=A-510
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart