ADV371426235 AChemBlock

2-Oxa-6-aza-spiro[3.3]heptane oxalic acid salt

  • MDL number MFCD11976178
  • Cas Number : 1045709-32-7
  • Molecular Weight : 189.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white powder
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-oxa-6-aza-spiro-3-3-heptane-oxalic-acid-salt.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=A-800
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart