ADV371426240 AChemBlock

Ethyl pyrazolo[1,5-a]pyrimidine-3-carboxylate

  • MDL number MFCD00847745
  • Cas Number : 115932-00-8
  • Molecular Weight : 191.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form yellow powder
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/ethyl-pyrazolo-1-5-a-pyrimidine-3-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=A-806
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart