ADV371426243 AChemBlock

3-Aminopyrazole

  • MDL number MFCD00005236
  • Cas Number : 1820-80-0
  • Molecular Weight : 83.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white to light yellow crystals
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/3-aminopyrazole.html

Safety Information

signalword Danger
pcodes P260;P264;P270;P280;P301+312;P301+330+331;P303+361+353;P304+340;P305+351+338;P310;P363
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=A-809
pictograms GHS05; GHS07
hstatements H302;H314
Availability details for
Quantity
  • Availability
  • Add to Cart