ADV371426244 AChemBlock

6-Bromopyrazolo[1,5-a]pyrimidine

  • MDL number MFCD09832894
  • Cas Number : 705263-10-1
  • Molecular Weight : 198.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form yellow solid/pale yellow solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/6-bromopyrazolo-1-5-a-pyrimidine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=A-810
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart