ADV371426248 AChemBlock

3,6-dibromopyrazolo[1,5-a]pyrimidine

  • MDL number MFCD12923112
  • Cas Number : 55405-68-0
  • Molecular Weight : 276.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/pyrazolo-1-5-a-pyrimidine-3-6-dibromo.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=A-814
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart