ADV371426264 AChemBlock

tert-butyl 3-hydroxy-3-methylpiperidine-1-carboxylate

  • MDL number MFCD13194152
  • Cas Number : 1104083-27-3
  • Molecular Weight : 215.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form colorless liquid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-piperidinecarboxylic-acid-3-hydroxy-3-methyl-1-1-dimethylethyl-ester.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=A-830
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart