ADV371426267 AChemBlock

2-Methyl-tetrahydro-pyran-4-one

  • MDL number MFCD15071428
  • Cas Number : 1193-20-0
  • Molecular Weight : 114.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Physical Form pale-yellow liquid
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/2-methyl-tetrahydro-pyran-4-one.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=A-836
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart