ADV371426268 AChemBlock

5-(Trifluoromethyl)-1H-pyrazol-3-amine

  • MDL number MFCD09834152
  • Cas Number : 852443-61-9
  • Molecular Weight : 151.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-trifluoromethyl-1h-pyrazol-3-amine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=A-839
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart