ADV371426271 AChemBlock

6-Hydroxy-2-aza-spiro[3.3]heptane-2-carboxylic acid tert-butyl ester

  • MDL number MFCD15071431
  • Cas Number : 1147557-97-8
  • Molecular Weight : 213.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/6-hydroxy-2-aza-spiro-3-3-heptane-2-carboxylic-acid-tert-butyl-ester.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=A-842
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart