ADV371426272 AChemBlock

6-Amino-2-aza-spiro[3.3]heptane-2-carboxylic acid tert-butyl ester

  • MDL number MFCD15071432
  • Cas Number : 1211586-09-2
  • Molecular Weight : 212.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/6-amino-2-aza-spiro-3-3-heptane-2-carboxylic-acid-tert-butyl-ester.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=A-844
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart