ADV371426273 AChemBlock

3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolane)-pyrazole

  • MDL number MFCD07368044
  • Cas Number : 844501-71-9
  • Molecular Weight : 194.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form white powder
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/3-4-4-5-5-tetramethyl-1-3-2-dioxaborolane-pyrazole.html

Safety Information

signalword warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=A-845
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart