ADV371426311 AChemBlock

8-Bromo-6-chloro-9-(tetrahydro-pyran-2-yl)-9H-purine

  • MDL number MFCD22124079
  • Cas Number : 218431-08-4
  • Molecular Weight : 317.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Room Temp
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.achemblock.com/8-bromo-6-chloro-9-tetrahydro-pyran-2-yl-9h-purine.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=A-888
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart