ADV371426312 AChemBlock

4,6-Dichloro-1H-pyrazolo[3,4-d]pyrimidine

  • MDL number MFCD09835493
  • Cas Number : 42754-96-1
  • Molecular Weight : 189

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white to beige to brown powder
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-6-dichloro-1h-pyrazolo-3-4-d-pyrimidine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=A-889
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart