ADV371426328 AChemBlock

[1,8]Naphthyridine-2-carbaldehyde

  • MDL number MFCD04971934
  • Cas Number : 64379-45-9
  • Molecular Weight : 158.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Store at 0-8 �C
Physical Form brown solid/pale brown solid
Shipping Temperature Store at 0-8 �C
Country of Origin CN
Product Link https://www.achemblock.com/1-8-naphthyridine-2-carbaldehyde.html

Safety Information

signalword Warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=A-950
pictograms GHS07
hstatements H302;H313;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart