ADV371426339 AChemBlock

3-(tert-Butoxycarbonylamino)cyclobutanecarboxylic acid

  • MDL number MFCD09864181
  • Cas Number : 946152-72-3
  • Molecular Weight : 215.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-tert-butoxycarbonylamino-cyclobutanecarboxylic-acid.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=A-969
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart