ADV371426343 AChemBlock

(2S)-1-[(tert-butoxy)carbonyl]-4,4-dimethylpyrrolidine-2-carboxylic acid

  • MDL number MFCD11040193
  • Cas Number : 1001353-87-2
  • Molecular Weight : 243.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/s-1-tert-butoxycarbonyl-4-4-dimethylpyrrolidine-2-carboxylic-acid.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=A-975
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart