ADV371426353 AChemBlock

(R)-Morpholine-2-carboxylic acid hydrochloride

  • MDL number MFCD13194711
  • Cas Number : 1273577-14-2
  • Molecular Weight : 167.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/r-morpholine-2-carboxylic-acid-hydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=B-1028
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart