ADV371426364 AChemBlock

6-Chloro-5-fluoroindolin-2-one

  • MDL number MFCD09839325
  • Cas Number : 100487-74-9
  • Molecular Weight : 185.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/6-chloro-5-fluoroindolin-2-one.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=B-1040
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart