ADV371426384 AChemBlock

(R)-2-Azetidinemethanol

  • MDL number MFCD10565789
  • Cas Number : 209329-11-3
  • Molecular Weight : 87.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form colorless liquid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/r-2-azetidinemethanol.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=B-1105
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart