ADV371426385 AChemBlock

(S)-2-Azetidinemethanol

  • MDL number MFCD10565790
  • Cas Number : 104587-62-4
  • Molecular Weight : 87.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form colorless liquid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/s-2-azetidinemethanol.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=B-1106
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart