ADV371426393 AChemBlock

6,7-dihydro-5H-Pyrrolo[3,4-b]pyridine dihydrochloride

  • MDL number MFCD09607967
  • Cas Number : 147740-02-1
  • Molecular Weight : 156.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form light-brown solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/6-7-dihydro-5h-pyrrolo-3-4-b-pyridine-dihydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=B-1200
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart