ADV371426395 AChemBlock

4-Bromopyridine-2-carboxylic acid

  • MDL number MFCD03426665
  • Cas Number : 30766-03-1
  • Molecular Weight : 202.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form brown solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-bromopyridine-2-carboxylic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=B-1209
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart